2-(2-Methylphenyl)ethanol 98%

BLDPBD77327-1GEA 9 EUR
BLDPBD77327-1G BLDPBD77327-100G BLDPBD77327-25G BLDPBD77327-10G BLDPBD77327-0.025G BLDPBD77327-5G
2-(2-Methylphenyl)ethanol 98%
2-(2-Methylphenyl)ethanol

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₂O
MW: 136,1900 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00044241
CAS-nummer: 19819-98-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR