(4-Methylmorpholin-2-yl)methanamine 95%

BLDPBD77434-5GEA 533 EUR
BLDPBD77434-5G BLDPBD77434-100MG BLDPBD77434-1G BLDPBD77434-250MG
(4-Methylmorpholin-2-yl)methanamine 95%
(4-Methylmorpholin-2-yl)methanamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₆H₁₄N₂O
MW: 130,1882 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD03990582
CAS-nummer: 141814-57-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR