1,2,4-Trihydroxybenzene 95%

BLDPBD7864-25GEA 31 EUR
BLDPBD7864-25G BLDPBD7864-5G BLDPBD7864-10G
1,2,4-Trihydroxybenzene 95%
1,2,4-Trihydroxybenzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₆O₃
MW: 126,11 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00002198
CAS-nummer: 533-73-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR