2-Chloro-5-cyanophenylboronic acid 98%

BLDPBD8029-1GEA 37 EUR
BLDPBD8029-1G BLDPBD8029-5G BLDPBD8029-250MG BLDPBD8029-25G
2-Chloro-5-cyanophenylboronic acid 98%
2-Chloro-5-cyanophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formule: C₇H₅BClNO₂
MW: 181,3841 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD06656271
CAS-nummer: 936249-33-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR