4-Bromomethyl-2-cyanobiphenyl 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD8054-10GEA 7 EUR
BLDPBD8054-10G BLDPBD8054-5G BLDPBD8054-25G
4-Bromomethyl-2-cyanobiphenyl 95%
4-Bromomethyl-2-cyanobiphenyl

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₄H₁₀BrN
MW: 272,14 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00671503
CAS-nummer: 114772-54-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR