3-(4-Chlorophenoxy)propionic acid 97%

BLDPBD80847-1GEA 23 EUR
BLDPBD80847-1G BLDPBD80847-250MG BLDPBD80847-5G
3-(4-Chlorophenoxy)propionic acid 97%
3-(4-Chlorophenoxy)propionic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₉ClO₃
MW: 200,6190 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00043504
CAS-nummer: 3284-79-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR