1-(3-(Trifluoromethyl)phenyl)thiourea 98%

BLDPBD81085-5GEA 18 EUR
BLDPBD81085-5G BLDPBD81085-1G
1-(3-(Trifluoromethyl)phenyl)thiourea 98%
1-(3-(Trifluoromethyl)phenyl)thiourea

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₇F₃N₂S
MW: 220,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00041171
CAS-nummer: 1736-70-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR