3,3-Dimethoxybutan-2-one 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD81346-5GEA 33 EUR
BLDPBD81346-5G BLDPBD81346-10G BLDPBD81346-1G
3,3-Dimethoxybutan-2-one 97%
3,3-Dimethoxybutan-2-one

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₁₂O₃
MW: 132,159 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00075266
CAS-nummer: 21983-72-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR