3-Bromo-2-methylpyridine 98%

BLDPBD8150-1GEA 6 EUR
BLDPBD8150-1G BLDPBD8150-10G BLDPBD8150-0.025G BLDPBD8150-5G BLDPBD8150-25G BLDPBD8150-500G BLDPBD8150-100G
3-Bromo-2-methylpyridine 98%
3-Bromo-2-methylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₆BrN
MW: 172,02 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00191224
CAS-nummer: 38749-79-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR