3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid 95%

BLDPBD81958-5GEA 765 EUR
BLDPBD81958-5G BLDPBD81958-100MG BLDPBD81958-250MG BLDPBD81958-1G
3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid 95%
3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C9H7NO3S
MW: 209,2200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00456558
CAS-nummer: 272437-84-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR