3,4-Dimethylthiophene 98%

BLDPBD82166-5GEA 269 EUR
BLDPBD82166-5G BLDPBD82166-1G BLDPBD82166-25G
3,4-Dimethylthiophene 98%
3,4-Dimethylthiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₆H₈S
MW: 112,1900 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01764731
CAS-nummer: 632-15-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR