3,5-Dichloro-4-methylaniline 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD82521-250MGEA 13 EUR
BLDPBD82521-250MG
3,5-Dichloro-4-methylaniline 97%
3,5-Dichloro-4-methylaniline
Formule: C7H7Cl2N
MW: 176,0400 g/mol
CAS-nummer: 54730-35-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR