3,5-Dihydroxybenzonitrile 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD82686-250MGEA 9 EUR
BLDPBD82686-250MG BLDPBD82686-1G BLDPBD82686-5G BLDPBD82686-25G
3,5-Dihydroxybenzonitrile 98%
3,5-Dihydroxybenzonitrile

Storage condition: Sealed in dry, 2-8°C

Formule: C₇H₅NO₂
MW: 135,12 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00016453
CAS-nummer: 19179-36-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR