2-Methylquinolin-3-amine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD83805-5GEA 620 EUR
BLDPBD83805-5G BLDPBD83805-100MG BLDPBD83805-250MG BLDPBD83805-1G
2-Methylquinolin-3-amine 97%
2-Methylquinolin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₀H₁₀N₂
MW: 158,1998 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08705651
CAS-nummer: 21352-22-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR