3-Aminothiophenol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD84603-25GEA 88 EUR
BLDPBD84603-25G BLDPBD84603-5G
3-Aminothiophenol 98%
3-Aminothiophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₇NS
MW: 125,19 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00007791
CAS-nummer: 22948-02-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR