6-Hydroxy-3,4-dihydroquinolin-2(1H)-one 98%

BLDPBD8462-500GEA 240 EUR
BLDPBD8462-500G BLDPBD8462-10G BLDPBD8462-0.5G BLDPBD8462-5G BLDPBD8462-25G BLDPBD8462-100G
6-Hydroxy-3,4-dihydroquinolin-2(1H)-one 98%
6-Hydroxy-3,4-dihydroquinolin-2(1H)-one

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₉NO₂
MW: 163,1700 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02179410
CAS-nummer: 54197-66-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR