3-Bromo-4-chlorophenol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD85025-100GEA 359 EUR
BLDPBD85025-100G BLDPBD85025-25G BLDPBD85025-1G BLDPBD85025-5G BLDPBD85025-10G
3-Bromo-4-chlorophenol 98%
3-Bromo-4-chlorophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₄BrClO
MW: 207,4500 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00070740
CAS-nummer: 13659-24-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR