N-ɑ-Boc-L-asparagine 98%

BLDPBD8506-100GEA 43 EUR
BLDPBD8506-100G BLDPBD8506-1000G BLDPBD8506-500G BLDPBD8506-25G BLDPBD8506-10G
N-ɑ-Boc-L-asparagine 98%
N-alpha-Boc-L-asparagine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₉H₁₆N₂O₅
MW: 232,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00038152
CAS-nummer: 7536-55-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR