Boc-Glu-OH 95%

BLDPBD8517-5GEA 7 EUR
BLDPBD8517-5G BLDPBD8517-10G BLDPBD8517-25G BLDPBD8517-100G
Boc-Glu-OH 95%
Boc-Glu-OH

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₀H₁₇NO₆
MW: 247,25 g/mol
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 2419-94-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR