(3-Chloro-5-fluorophenyl)acetic acid 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD85932-5GEA 78 EUR
BLDPBD85932-5G BLDPBD85932-25G BLDPBD85932-100G BLDPBD85932-250MG BLDPBD85932-1G
(3-Chloro-5-fluorophenyl)acetic acid 95%
(3-Chloro-5-fluorophenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formule: (C₆H₃ClF)CH₂COOH
MW: 188,59 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03788475
CAS-nummer: 202001-00-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR