Methyl-3-(hydroxymethyl)benzoate 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD87357-250MGEA 55 EUR
BLDPBD87357-250MG BLDPBD87357-1G BLDPBD87357-100MG
Methyl-3-(hydroxymethyl)benzoate 95%
Methyl-3-(hydroxymethyl)benzoate

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₀O₃
MW: 166,1739 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06202752
CAS-nummer: 67853-03-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR