3-Methyl-5-phenylisoxazole-4-carboxylic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD88125-250MGEA 24 EUR
BLDPBD88125-250MG BLDPBD88125-10G BLDPBD88125-1G BLDPBD88125-5G
3-Methyl-5-phenylisoxazole-4-carboxylic acid 97%
3-Methyl-5-phenylisoxazole-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C11H9NO3
MW: 203,1941 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03086120
CAS-nummer: 17153-21-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR