Methyl-3-(phenylamino)propanoate 98%

BLDPBD88897-1GEA 73 EUR
BLDPBD88897-1G BLDPBD88897-250MG
Methyl-3-(phenylamino)propanoate 98%
Methyl-3-(phenylamino)propanoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₁₀H₁₃NO₂
MW: 179,2200 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00089334
CAS-nummer: 21911-84-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR