4-Imidazol-1-ylmethylphenylamine 98%

BLDPBD89527-250MGEA 21 EUR
BLDPBD89527-250MG BLDPBD89527-5G BLDPBD89527-1G BLDPBD89527-25G
4-Imidazol-1-ylmethylphenylamine 98%
4-Imidazol-1-ylmethylphenylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₀H₁₁N₃
MW: 173,2144 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06660785
CAS-nummer: 56643-85-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR