4-(4-Bromophenylsulphonyl)morpholine 98%

BLDPBD90413-1GEA 31 EUR
BLDPBD90413-1G BLDPBD90413-25G BLDPBD90413-5G BLDPBD90413-0.025G
4-(4-Bromophenylsulphonyl)morpholine 98%
4-(4-Bromophenylsulphonyl)morpholine

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₁₂BrNO₃S
MW: 306,1800 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00194739
CAS-nummer: 834-67-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR