4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid 98%

BLDPBD92402-25GEA 765 EUR
BLDPBD92402-25G BLDPBD92402-5G BLDPBD92402-1G BLDPBD92402-100MG BLDPBD92402-250MG BLDPBD92402-10G
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid 98%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₉H₁₀O₂S
MW: 182,24 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00464469
CAS-nummer: 40133-07-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR