4,6-Dichloropicolinic acid 98%

BLDPBD92790-1GEA 7 EUR
BLDPBD92790-1G BLDPBD92790-5G BLDPBD92790-250MG BLDPBD92790-10G BLDPBD92790-25G BLDPBD92790-100G
4,6-Dichloropicolinic acid 98%
4,6-Dichloropicolinic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₃Cl₂NO₂
MW: 192,0000 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09910267
CAS-nummer: 88912-25-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR