4-Amino-1,2,5-oxadiazole-3-carboxylic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD93531-5GEA 31 EUR
BLDPBD93531-5G BLDPBD93531-250MG BLDPBD93531-25G BLDPBD93531-1G BLDPBD93531-100G
4-Amino-1,2,5-oxadiazole-3-carboxylic acid 97%
4-Amino-1,2,5-oxadiazole-3-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formule: C₃H₃N₃O₃
MW: 129,0742 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD00235172
CAS-nummer: 78350-50-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR