(4-Amino-2-methyl-5-pyrimidinyl)methanol 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD93954-250MGEA 47 EUR
BLDPBD93954-250MG BLDPBD93954-5G BLDPBD93954-1G
(4-Amino-2-methyl-5-pyrimidinyl)methanol 95%
(4-Amino-2-methyl-5-pyrimidinyl)methanol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₉N₃O
MW: 139,1552 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00849239
CAS-nummer: 73-67-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR