4-Bromoisoindolin-1-one 97%

BLDPBD95248-0.025GEA 0 EUR
BLDPBD95248-0.025G BLDPBD95248-25G BLDPBD95248-5G BLDPBD95248-10G BLDPBD95248-100G BLDPBD95248-1G BLDPBD95248-250MG
4-Bromoisoindolin-1-one 97%
4-Bromoisoindolin-1-one

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₆BrNO
MW: 212,0400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09701286
CAS-nummer: 337536-15-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR