4-Bromophthalonitrile 96%

Leverancier: BLD PHARMATECH GMBH

BLDPBD95351-100MGEA 22 EUR
BLDPBD95351-100MG BLDPBD95351-25G BLDPBD95351-1G BLDPBD95351-250MG BLDPBD95351-5G
4-Bromophthalonitrile 96%
4-Bromophthalonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₃BrN₂
MW: 207,0300 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00221794
CAS-nummer: 70484-01-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR