4-Bromo-1H-pyrrole-2-carboxylic acid 95%

BLDPBD95366-250MGEA 7 EUR
BLDPBD95366-250MG BLDPBD95366-10G BLDPBD95366-25G BLDPBD95366-100G BLDPBD95366-1G BLDPBD95366-5G
4-Bromo-1H-pyrrole-2-carboxylic acid 95%
4-Bromo-1H-pyrrole-2-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₅H₄BrNO₂
MW: 189,9948 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00832862
CAS-nummer: 27746-02-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR