4,6-Dichloro-2-(methylthio)pyrimidine 98%

BLDPBD9544-25GEA 10 EUR
BLDPBD9544-25G BLDPBD9544-10G
4,6-Dichloro-2-(methylthio)pyrimidine 98%
4,6-Dichloro-2-(methylthio)pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formule: C₅H₄Cl₂N₂S
MW: 195,07 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00006086
CAS-nummer: 6299-25-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR