4-Methylthiazole-5-carboxylic acid 98%

BLDPBD9584-25GEA 59 EUR
BLDPBD9584-25G BLDPBD9584-5G BLDPBD9584-100G BLDPBD9584-500G BLDPBD9584-10G BLDPBD9584-1G
4-Methylthiazole-5-carboxylic acid 98%
4-Methylthiazole-5-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₅H₅NO₂S
MW: 143,17 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00626872
CAS-nummer: 20485-41-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR