1H-Pyrazole-4-carbonitrile 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD96662-100MGEA 7 EUR
BLDPBD96662-100MG BLDPBD96662-10G BLDPBD96662-5G BLDPBD96662-100G BLDPBD96662-1G BLDPBD96662-250MG BLDPBD96662-25G
1H-Pyrazole-4-carbonitrile 98%
1H-Pyrazole-4-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₄H₃N₃
MW: 93,0900 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00040250
CAS-nummer: 31108-57-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR