3,3-Dimethylbutyric acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD9678-10GEA 6 EUR
BLDPBD9678-10G BLDPBD9678-25G
3,3-Dimethylbutyric acid 98%
3,3-Dimethylbutyric acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₆H₁₂O₂
MW: 116,16 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00002715
CAS-nummer: 1070-83-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR