(1S,2S)-(+)-1,2-Diaminocyclohexane 96%

Leverancier: BLD PHARMATECH GMBH

BLDPBD9696-1GEA 6 EUR
BLDPBD9696-1G BLDPBD9696-10G BLDPBD9696-25G BLDPBD9696-5G
(1S,2S)-(+)-1,2-Diaminocyclohexane 96%
(1S,2S)-(+)-1,2-Diaminocyclohexane

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₁₄N₂
MW: 114,19 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00062986
CAS-nummer: 21436-03-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR