4-Ethylphenyl isocyanate 98%

BLDPBD97024-250MGEA 14 EUR
BLDPBD97024-250MG
4-Ethylphenyl isocyanate 98%
4-Ethylphenyl isocyanate

Storage condition: Inert atmosphere, 2-8°C

Formule: C₉H₉NO
MW: 147,18 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00013881
CAS-nummer: 23138-50-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR