1-Broom-2,3,5-trifluorobenzeen 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD9719-25GEA 110 EUR
BLDPBD9719-25G BLDPBD9719-1G BLDPBD9719-5G
1-Broom-2,3,5-trifluorobenzeen 98%
1-Broom-2,3,5-trifluorobenzeen

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₆H₂BrF₃
MW: 210,98 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00012232
CAS-nummer: 133739-70-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR