4-Fluoroisatin 95%

BLDPBD97329-250MGEA 59 EUR
BLDPBD97329-250MG BLDPBD97329-1G BLDPBD97329-100MG BLDPBD97329-5G
4-Fluoroisatin 95%
4-Fluoroisatin

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₄FNO₂
MW: 165,1200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01175824
CAS-nummer: 346-34-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR