4-Fluoro-N-isopropylaniline 96%

BLDPBD97344-250MGEA 42 EUR
BLDPBD97344-250MG BLDPBD97344-1G BLDPBD97344-5G
4-Fluoro-N-isopropylaniline 96%
4-Fluoro-N-isopropylaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₉H₁₂FN
MW: 153,2000 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00270760
CAS-nummer: 70441-63-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR