2-Bromo-4,6-difluorophenol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD9745-25GEA 199 EUR
BLDPBD9745-25G BLDPBD9745-1G BLDPBD9745-5G BLDPBD9745-100G
2-Bromo-4,6-difluorophenol 98%
2-Bromo-4,6-difluorophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₃BrF₂O
MW: 208,99 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00070752
CAS-nummer: 98130-56-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR