4'-Methoxyacetophenone 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD98473-500GEA 24 EUR
BLDPBD98473-500G BLDPBD98473-25G BLDPBD98473-0.025G BLDPBD98473-1000G BLDPBD98473-100G
4'-Methoxyacetophenone 95%
4'-Methoxyacetophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₉H₁₀O₂
MW: 150,18 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00008745
CAS-nummer: 100-06-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR