4-Methoxyazobenzene 98%
|
|
BLDPBD98478-1GEA
40
EUR
BLDPBD98478-1G
4-Methoxyazobenzene 98%
4-Methoxyazobenzene
Storage condition: Sealed in dry, Room Temperature
Formule:
C₁₃H₁₂N₂O MW: 212,2472 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00039804 CAS-nummer: 2396-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...