4-Methylbenzimidazole 98%

BLDPBD98694-1GEA 117 EUR
BLDPBD98694-1G BLDPBD98694-100MG BLDPBD98694-250MG BLDPBD98694-5G
4-Methylbenzimidazole 98%
4-Methylbenzimidazole

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₈N₂
MW: 132,1600 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09842603
CAS-nummer: 4887-83-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR