4-Octylaniline 98%

BLDPBD99631-25GEA 140 EUR
BLDPBD99631-25G BLDPBD99631-5G BLDPBD99631-1G
4-Octylaniline 98%
4-Octylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₄H₂₃N
MW: 205,34 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00007929
CAS-nummer: 16245-79-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR