4-Phenyl-1-butyne 98%

BLDPBD99918-25GEA 521 EUR
BLDPBD99918-25G BLDPBD99918-1G BLDPBD99918-250MG BLDPBD99918-5G
4-Phenyl-1-butyne 98%
4-Phenyl-1-butyne

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₁₀
MW: 130,19 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00041667
CAS-nummer: 16520-62-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR