p-Terphenyl 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD99965-25GEA 10 EUR
BLDPBD99965-25G
p-Terphenyl 98%
p-Terphenyl

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₈H₁₄
MW: 230,31 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003061
CAS-nummer: 92-94-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR