L(+)-Glutamic acid

Leverancier: EDQM

EDQMG0355000EA 208 EUR
EDQMG0355000
L(+)-Glutamic acid
L(+)-Glutamic acid
Formule: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147,13 g/mol
Kookpunt: 325 °C (1013 hPa)
Smeltpunt: 160 °C
Dichtheid: 1,538 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00002634
CAS-nummer: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://be.vwr.com/stibo/web/std.lang.all/98/27/2299827.jpg

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR