Germanium(IV) oxide
Leverancier: VWR Chemicals
|
Waarschuwing
|
24362.104EA
0
EUR
24362.104
Germanium(IV) oxide
Germanium(IV) oxide
Formule:
GeO₂ MW: 104,61 g/mol Smeltpunt: 1110 °C Dichtheid: 3,64 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00011030 CAS-nummer: 1310-53-8 EINECS: 215-180-8 Merck Index: 12,04417 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...